Learn More
XY 018, Tocris Bioscience™
GREENER_CHOICE
ROR/NR1F Antagonists
Marca:  Tocris Bioscience 6223/50
Encompass
Código de producto. 15937920
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos »
Descripción
Descripción
XY 018 is a RORγ antagonist (EC50 = 190 nM). Suppresses androgen receptor expression in prostate cancer cell lines. Also inhibits growth of tumors in androgen receptor-positive xenograft models.
Especificaciones
Especificaciones
1873358-87-2
N-[2′-Fluoro-4′-[2,2,2-trifluoro-1-hydroxy-1-(trifluoromethyl)ethyl][1,1′biphenyl]-4-yl]-2-nitrobenzeneacetamide