Learn More
XL413 hydrochloride, MedChemExpress
Código de producto. 16476408
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16476408 10 mg 10 miligramos
16468997 5 mg 5 miligramos
16452517 50 mg 50 miligramos
16496696 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16476408 Proveedor Medchem Express N.º de proveedor HY15260A
MedChemExpress XL413 (BMS-863233) hydrochloride is a potent, selective and ATP competitive inhibitor of Cdc7, with an IC50 of 3.4 nM, and also shows potent effect with IC50s of 215, 42 nM on CK2, PIM1, respectively, and an EC50 of 118 nM on pMCM.
XL413 (BMS-863233) hydrochloride is a potent, selective and ATP competitive inhibitor of Cdc7, with an IC50 of 3.4 nM, and also shows potent effect with IC50s of 215, 42 nM on CK2, PIM1, respectively, and an EC50 of 118 nM on pMCM.
Especificaciones
Nombre del producto químico o material XL413 hydrochloride
Forma física Solid
CAS 2062200-97-7
Color Blanco
Cantidad 10 mg
Notas de grado de pureza Research
Fórmula molecular C14H13Cl2N3O2
Para utilizar con (aplicación) Cancer-Kinase/protease
Sinónimo BMS-863233 hydrochloride
Información de solubilidad H2O : 10 mg/mL (30.66 mM; Need ultrasonic) ∣DMSO : 3.4 mg/mL (10.42 mM; Need ultrasonic and warming)
SMILES O=C1C(OC2=CC=C(Cl)C=C23)=C3N=C([C@H]4NCCC4)N1.[H]Cl
Peso molecular (g/mol) 326.18
Formula Weight (peso de la fórmula) 326.18
Porcentaje de pureza 99.82%