Learn More
Tocris Bioscience™ WM 8014
Descripción
WM 8014 is a potent and selective lysine acetyltransferase KAT6A and KAT6B inhibitor (IC50 values are 8 and 28 nM, respectively). Exhibits > 10-fold selectivity for KAT6A/B over KAT7 and KAT5, and no significant activity against KAT8, KAT2A/2B, and KAT3A/B. Induces cell cycle arrest in G1/G0 in MEFs and inhibits proliferation of lymphoma cells in vitro. Cell permeant. Suitable for use in vitro.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N'-(4-Fluoro-5-methyl-[1,1 '-biphenyl]-3-carbonyl)benzenesulfonohydrazide |
| CAS | 2055397-18-5 |
| Cantidad | 10 mg |
| Objetivo | Histone Acetyltransferase Inhibitors |
| Fórmula molecular | C20H17FN2O3S |
| Pureza | 0.98 |