Learn More
Tocris Bioscience™ WM 3835
Descripción
WM 3835 is a lysine acetyltransferase HBO1 (KAT7) inhibitor. Reduces H3K14ac levels and inhibits growth and viability of acyte myeloid leukemia cell lines in vitro. Cell-permeable.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N'-(4-Fluoro-5-methyl-[1,1 '-biphenyl]-3-carbonyl)-3-hydroxybenzenesulfonohydrazide |
| CAS | 2229025-70-9 |
| Cantidad | 50 mg |
| Objetivo | Histone Acetyltransferase Inhibitors |
| Fórmula molecular | C20H17FN2O4S |
| Pureza | 0.98 |