Learn More
Tocris Bioscience™ WAY 181187 oxalate
Descripción
WAY 181187 oxalate is a high affinity and selective 5-HT6 agonist (Ki = 2.2 nM). Displays 60-fold selectivity over other 5-HT and monoamine receptors. Increases firing rate of 5-HT neurons. Modifies sleep and wakefulness profiles following direct injections into the dorsal raphe nucleus, in vivo. Orally bioavailable.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 1-[(6-Chloroimidazo[2,1-b]thiazol-5-yl)sulfonyl]-1H-indole-3-ethanamine oxalate |
| CAS | 1883548-85-3 |
| Cantidad | 50 mg |
| Objetivo | 5-HT6 Receptor Agonists |
| Fórmula molecular | C15H13ClN4O2S2.C2H2O4 |
| Formula Weight (peso de la fórmula) | 470.91 |
| Pureza | >98% |