Learn More
WAY-151932, MedChemExpress
Código de producto. 16467758
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
1 mg
5 mg
10 mg
Tamaño de la unidad:
1 miligramo
10 miligramos
5 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16467758 10 mg 10 miligramos
16431809 1 mg 1 miligramo
16400228 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 16467758 missing translation for 'mfr' Medchem Express missing translation for 'supplierNo' HY19381
This item is not returnable. View return policy
MedChemExpress WAY-151932 is a vasopressin V2-receptor agonist with IC50 of 80.3 nM and 778 nM in human-V2 binding and V1a binding assay.
WAY-151932 is a vasopressin V2-receptor agonist with IC50 of 80.3 nM and 778 nM in human-V2 binding and V1a binding assay.
Specifications
Nombre del producto químico o material WAY-151932
CAS 220460-92-4
Color Off-White
Notas de grado de pureza Research
Fórmula molecular C23H19ClN4O
Cantidad 10 mg
Sinónimo VNA-932 WAY-VNA 932
Información de solubilidad DMSO : 200 mg/mL (496.43 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(N2N=C(C)C=C2)C=C1Cl)N3CC4=CC=CN4CC5=CC=CC=C53
Peso molecular (g/mol) 402.88
Formula Weight (peso de la fórmula) 402.88
Porcentaje de pureza 99.44%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Metabolism-protein/nucleotide metabolism