Learn More
W-13 hydrochloride, Tocris Bioscience™
Código de producto. 30001084
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
30001084 10 mg 10 miligramos
18044682 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 30001084 Proveedor Tocris Bioscience N.º de proveedor 0361/10
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos
Calmodulin antagonist
Calmodulin antagonist (inhibits calmodulin activated PDE activity with an IC50 of 68 μM). Inhibits growth of tamoxifen-resistant breast cancer cells.
TRUSTED_SUSTAINABILITY
CAS 88519-57-7
Fórmula molecular C14H18Cl2N2O2S
Peso molecular (g/mol) 349.27
Clave InChI QKAALLVQBOLELJ-UHFFFAOYSA-N
Sinónimo w-13 hydrochloride, n-4-aminobutyl-5-chloronaphthalene-2-sulfonamide hydrochloride, w-13, hydrochloride, n-4-aminobutyl-5-chloro-2-naphthalenesulfonamide hydrochloride, 2-naphthalenesulfonamide,n-4-aminobutyl-5-chloro-, hydrochloride 1:1, n-4-aminobutyl-5-chloro-2-naphthalenesulfonamide, hcl, acmc-209qtx, n-4-aminobutyl-5-chloro-2-naphthalenesulfonamide hcl, n-4-aminobutyl-5-chloronaphthalene-2-sulfonamidehydrochloride, n-4-aminobutyl-5-chloro-2-naphthalene sulfonamide hydro chloride
PubChem CID 16760706
ChEBI CHEBI:75367
Nombre IUPAC N-(4-aminobutyl)-5-chloronaphthalene-2-sulfonamide;hydrochloride
SMILES C1=CC2=C(C=CC(=C2)S(=O)(=O)NCCCCN)C(=C1)Cl.Cl
Especificaciones
Nombre del producto químico o material W-13 hydrochloride
Cantidad 10 mg
Formula Weight (peso de la fórmula) 349.27
Porcentaje de pureza >98%