Learn More
VU591, MedChemExpress
Código de producto. 16474208
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
50 mg
Tamaño de la unidad:
10 miligramos
25 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16474208 10 mg 10 miligramos
16456927 5 mg 5 miligramos
16432996 25 mg 25 miligramos
16410557 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16474208 Proveedor Medchem Express N.º de proveedor HY108585A
MedChemExpress VU591 is a potent, selective renal outer medullary potassium channel (ROMK or Kir1.1) inhibitor, with an IC50 of 0.24 μM. VU591 can be used for neurological research with HY-108585 (the equivalent of VU591 hydrochloride).
VU591 is a potent, selective renal outer medullary potassium channel (ROMK or Kir1.1) inhibitor, with an IC50 of 0.24 μM. VU591 can be used for neurological research with HY-108585 (the equivalent of VU591 hydrochloride).
Especificaciones
Nombre del producto químico o material VU591
Forma física Solid
CAS 1222810-74-3
Color Gray
Cantidad 10 mg
Notas de grado de pureza Research
Fórmula molecular C16H12N6O5
Información de solubilidad DMSO : 62.5 mg/mL (169.70 mM; Need ultrasonic)
SMILES O=[N+]([O-])C1=CC(NC(COCC2=NC3=C(N2)C=C([N+]([O-])=O)C=C3)=N4)=C4C=C1
Peso molecular (g/mol) 368.3
Formula Weight (peso de la fórmula) 368.3
Porcentaje de pureza 98.07%