Learn More
VU591 hydrochloride, MedChemExpress
Código de producto. 16446927
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
Tamaño de la unidad:
5 miligramos
16446927 5 mg 5 miligramos
1 options
Código de producto. 16446927 Proveedor Medchem Express N.º de proveedor HY108585
MedChemExpress VU591 hydrochloride is a potent, selective renal outer medullary potassium channel (ROMK or Kir1.1) inhibitor, with an IC50 of 0.24 μM. VU591 hydrochloride can be used for neurological research with HY-108585A (the equivalent of VU591 hydrochloride).
VU591 hydrochloride is a potent, selective renal outer medullary potassium channel (ROMK or Kir1.1) inhibitor, with an IC50 of 0.24 μM. VU591 hydrochloride can be used for neurological research with HY-108585A (the equivalent of VU591 hydrochloride).
Especificaciones
Nombre del producto químico o material VU591 hydrochloride
Forma física Solid
CAS 1315380-70-1
Color Gray
Cantidad 5 mg
Notas de grado de pureza Research
Fórmula molecular C16H13ClN6O5
Información de solubilidad DMSO : 16.67 mg/mL (41.18 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=[N+](C1=CC=C2N=C(COCC3=NC4=CC=C([N+]([O-])=O)C=C4N3)NC2=C1)[O-].[H]Cl
Peso molecular (g/mol) 404.76
Formula Weight (peso de la fórmula) 404.76
Porcentaje de pureza 98.01%