Learn More
Tocris Bioscience™ VU 0546110
Descripción
VU 0546110 is a selective inhibitor of the sperm-specific potassium channel SLO3 (IC50 = 64 μM). It shows 40-fold selectivity over SLO1. It completely blocks heterologous SLO3 currents and endogenous K+ currents. VU 0546110 inhibits sperm hyperpolarisation and prevents its hyperactivated mobility.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 1-[4-[[(4-Methoxyphenyl)thio]methyl]-1-piperidinyl]-2-[4-[(1-methylethyl)sulfonyl]phenyl]ethanone |
| CAS | 1421523-00-3 |
| Cantidad | 50 mg |
| Objetivo | Non-selective/Other Potassium Channel Blockers |
| Fórmula molecular | C24H31NO4S2 |
| Pureza | 0.98 |