Learn More
Voxtalisib, MedChemExpress
Código de producto. 16496908
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
25 miligramos
5 miligramos
50 miligramos
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
16496908 10 mg 10 miligramos
16492967 50 mg 50 miligramos
16483506 25 mg 25 miligramos
16446986 100 mg 100 miligramos
16429447 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Código de producto. 16496908 Proveedor Medchem Express N.º de proveedor HY15900
MedChemExpress Voxtalisib (XL765) is a potent PI3K inhibitor, which has a similar activity toward class I PI3K (IC50s=39, 113, 9 and 43 nM for p110α, p110β, p110γ and p110δ, respectively), also inhibits DNA-PK (IC50=150 nM) and mTOR (IC50=157 nM). Voxtalisib (XL765) inhibits mTORC1 and mTORC2 with IC50s of 160 and 910 nM, respectively.
Voxtalisib (XL765) is a potent PI3K inhibitor, which has a similar activity toward class I PI3K (IC50s=39, 113, 9 and 43 nM for p110α, p110β, p110γ and p110δ, respectively), also inhibits DNA-PK (IC50=150 nM) and mTOR (IC50=157 nM). Voxtalisib (XL765) inhibits mTORC1 and mTORC2 with IC50s of 160 and 910 nM, respectively.
Especificaciones
Nombre del producto químico o material Voxtalisib
Forma física Solid
CAS 934493-76-2
Color Marrón
Cantidad 10 mg
Notas de grado de pureza Research
Fórmula molecular C13H14N6O
Para utilizar con (aplicación) Cancer-Kinase/protease
Sinónimo XL765 SAR245409
Información de solubilidad DMSO : 10 mg/mL (37.00 mM; Need ultrasonic)
SMILES O=C1C(C2=NNC=C2)=CC3=C(C)N=C(N)N=C3N1CC
Peso molecular (g/mol) 270.29
Formula Weight (peso de la fórmula) 270.29
Porcentaje de pureza 98.68%