Learn More
Vorinostat, MedChemExpress
Código de producto. 16499845
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
250 mg
500 mg
1 g
5 g
Tamaño de la unidad:
1 g
250 miligramos
5 g
500 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16499845 250 mg 250 miligramos
16461546 500 mg 500 miligramos
16460979 1 g 1 g
16446457 5 g 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16499845 Proveedor Medchem Express N.º de proveedor HY10221
MedChemExpress Vorinostat (SAHA) is a potent and orally active pan-inhibitor of HDAC1, HDAC2 and HDAC3 (Class I), HDAC6 and HDAC7 (Class II) and HDAC11 (Class IV), with ID50 values of 10 nM and 20 nM for HDAC1 and HDAC3, respectively. Vorinostat induces cell apoptosis. Vorinostat is also an effective inhibitor of human papillomaviruse (HPV)-18 DNA amplification.
Vorinostat (SAHA) is a potent and orally active pan-inhibitor of HDAC1, HDAC2 and HDAC3 (Class I), HDAC6 and HDAC7 (Class II) and HDAC11 (Class IV), with ID50 values of 10 nM and 20 nM for HDAC1 and HDAC3, respectively. Vorinostat induces cell apoptosis. Vorinostat is also an effective inhibitor of human papillomaviruse (HPV)-18 DNA amplification.
Especificaciones
Nombre del producto químico o material Vorinostat
Forma física Solid
CAS 149647-78-9
Color Blanco
Cantidad 250 mg
Notas de grado de pureza Research
Fórmula molecular C14H20N2O3
Para utilizar con (aplicación) Cancer-programmed cell death
Sinónimo SAHA Suberoylanilide hydroxamic acid
Información de solubilidad DMSO : ≥ 100 mg/mL (378.33 mM)
SMILES O=C(NC1=CC=CC=C1)CCCCCCC(NO)=O
Peso molecular (g/mol) 264.32
Formula Weight (peso de la fórmula) 264.32
Porcentaje de pureza 95.0%