Learn More
Voriconazole, MedChemExpress
Código de producto. 16427946
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
50 mg
100 mg
Tamaño de la unidad:
100 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16427946 100 mg 100 miligramos
16404107 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16427946 Proveedor Medchem Express N.º de proveedor HY76200
MedChemExpress Voriconazole (UK-109496) is a second-generation, broad-spectrum triazole antifungal agent that inhibits fungal ergosterol biosynthesis. Voriconazole exerts its antifungal activity by inhibition of 14-α-lanosterol demethylation, which is mediated by fungal cytochrome P450 enzymes.
Voriconazole (UK-109496) is a second-generation, broad-spectrum triazole antifungal agent that inhibits fungal ergosterol biosynthesis. Voriconazole exerts its antifungal activity by inhibition of 14-α-lanosterol demethylation, which is mediated by fungal cytochrome P450 enzymes.
Especificaciones
Nombre del producto químico o material Voriconazole
Forma física Solid
CAS 137234-62-9
Color Blanco
Cantidad 100 mg
Notas de grado de pureza Research
Fórmula molecular C16H14F3N5O
Para utilizar con (aplicación) COVID-19-immunoregulation
Sinónimo UK-109496
Información de solubilidad DMSO : ≥ 50 mg/mL (143.14 mM) ∣H2O : 0.17 mg/mL (0.49 mM; Need ultrasonic)
SMILES O[C@@]([C@@H](C)C1=NC=NC=C1F)(CN2N=CN=C2)C(C(F)=C3)=CC=C3F
Peso molecular (g/mol) 349.31
Formula Weight (peso de la fórmula) 349.31
Porcentaje de pureza 99.75%