Learn More
Vorasidenib, MedChemExpress
Código de producto. 16483938
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
2 mg
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
2 miligramos
5 miligramos
50 miligramos
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
16483938 10 mg 10 miligramos
16480106 2 mg 2 miligramos
16466657 5 mg 5 miligramos
16450297 50 mg 50 miligramos
16424966 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Código de producto. 16483938 Proveedor Medchem Express N.º de proveedor HY104042
MedChemExpress Vorasidenib (AG-881) is an orally available, brain penetrant second-generation dual mutant isocitrate dehydrogenases 1 and 2 (mIDH1/2) inhibitor. Vorasidenib (AG-881) exhibits nanomolar inhibition of (D)-2-hydroxyglutarate (D-2-HG), and the IC50 ranges of 0.04∼22 nM against IDH1 R132C, IDH1 R132G, IDH1 R132H and IDH1 R132S and 7∼14 nM against IDH2 R140Q and 130 nM against IDH2 R172K.
Vorasidenib (AG-881) is an orally available, brain penetrant second-generation dual mutant isocitrate dehydrogenases 1 and 2 (mIDH1/2) inhibitor. Vorasidenib (AG-881) exhibits nanomolar inhibition of (D)-2-hydroxyglutarate (D-2-HG), and the IC50 ranges of 0.04∼22 nM against IDH1 R132C, IDH1 R132G, IDH1 R132H and IDH1 R132S and 7∼14 nM against IDH2 R140Q and 130 nM against IDH2 R172K.
Especificaciones
Nombre del producto químico o material Vorasidenib
CAS 1644545-52-7
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C14H13ClF6N6
Cantidad 10 mg
Sinónimo AG-881
Información de solubilidad DMSO : 100 mg/mL (241.11 mM; Need ultrasonic)
SMILES C[C@@H](NC1=NC(C2=NC(Cl)=CC=C2)=NC(N[C@H](C)C(F)(F)F)=N1)C(F)(F)F
Peso molecular (g/mol) 414.74
Formula Weight (peso de la fórmula) 414.74
Porcentaje de pureza 99.3%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death