Learn More
Volasertib, TRC
Product Code. 30451688
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Cantidad:
5 mg
10 mg
25 mg
missing translation for 'unitSize'
25 miligramos
5 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30451688 25 mg 25 miligramos
30451675 10 mg -
30452368 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30451688 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCV57518025MG
This item is not returnable. View return policy
Moléculas orgánicas de alta pureza y estándares analíticos, implementados estratégicamente en todo el mundo para impulsar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 755038-65-4
Fórmula molecular C34H50N8O3
Peso molecular (g/mol) 618.81
Sinónimo BI 6727, N-[trans-4-[4-(Cyclopropylmethyl)-1-piperazinyl]cyclohexyl]-4-[[(7R)-7-ethyl-5,6,7,8-tetrahydro-5-methyl-8-(1-methylethyl)-6-oxo-2-pteridinyl]amino]-3-methoxy-benzamide
Nombre IUPAC N-[4-[4-(ciclopropimetil)piperazina-1-il]ciclohexil]-4-[[(7R)-7-etil-5-metil-6-oxo-8-propano-2-il-7H-pteridina-2-il]amino]-3-metoxibenzamida
SMILES CC[C@H]1N(C(C)C)c2nc(Nc3ccc(cc3OC)C(=O)N[C@@H]4CC[C@H](CC4)N5CCN(CC6CC6)CC5)ncc2N(C)C1=O
Specifications
Nombre del producto químico o material Volasertib
Cantidad 25 mg
Fórmula InChI InChI=1S/C34H50N8O3/c1-6-28-33(44)39(4)29-20-35-34(38-31(29)42(28)22(2)3)37-27-14-9-24(19-30(27)45-5)32(43)36-25-10-12-26(13-11-25)41-17-15-40(16-18-41)21-23-7-8-23/h9,14,19-20,22-23,25-26,28H,6-8,10-13,15-18,21H2,1-5H3,(H,36,43)(H,35,37,38)/t25-,26-,28-/m1/s1
Formula Weight (peso de la fórmula) 618.4006