Learn More
Voglibose, MedChemExpress
Código de producto. 16494207
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
50 mg
100 mg
Tamaño de la unidad:
100 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16494207 50 mg 50 miligramos
16458066 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16494207 Proveedor Medchem Express N.º de proveedor HYB0025
MedChemExpress Voglibose is an N-substituted derivative of valiolamine, excellent inhibitory activity against α-glucosidases and its action against hyperglycemia and various disorders caused by hyperglycemia.
Voglibose is an N-substituted derivative of valiolamine, excellent inhibitory activity against α-glucosidases and its action against hyperglycemia and various disorders caused by hyperglycemia.
Especificaciones
Nombre del producto químico o material Voglibose
CAS 83480-29-9
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C10H21NO7
Cantidad 50 mg
Información de solubilidad H2O : 250 mg/mL (935.35 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (374.14 mM; Need ultrasonic)
SMILES O[C@@H]1[C@@](O)(CO)C[C@H](NC(CO)CO)[C@H](O)[C@H]1O
Peso molecular (g/mol) 267.28
Formula Weight (peso de la fórmula) 267.28
Porcentaje de pureza 98.0%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Metabolism-sugar/lipid metabolism