Learn More
Thermo Scientific Chemicals Clorhidrato de vitamina B12a
Código de producto. 16348297 Tienda Thermo Scientific Chemicals Productos
Cambiar vista
Click to view available options
Cantidad:
1 g
5 g
Tamaño de la unidad:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16348297 1 g 1 g
16358297 5 g 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16348297 Proveedor Thermo Scientific Chemicals N.º de proveedor 462080010
Solicitar formato a granel o personalizado
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS 58288-50-9
Fórmula molecular C62H91ClCoN13O15P
Peso molecular (g/mol) 1383.84
Clave InChI BBQILPBAMXKFOB-UHFFFAOYNA-L
Nombre IUPAC clorhidrato de λ²-cobalto(2+) 4,9,14-tris(2-carbamoyletil)-3,8,19-tris(carbamoilmetil)-18-{2-[(2-{[5-(5,6-dimetil-1H-1,3-benzodiazol-1-il)-4-hidroxi-2-(hidroximetil)oxolan-3-il fosfonato]oxi}propil)carbamoil]etil}-2,3,6,8,13,13,16,18-octametil-20,21,22,23-tetraazapentaciclo[15.2.1.1²,⁵.1⁷,¹⁰.1¹²,¹⁵]tricosa-5(23),6,10(22),11,15(21),16-hexaen-20-uro hidrato
SMILES O.Cl.[Co++].CC(CNC(=O)CCC1(C)C(CC(N)=O)C2[N-]C1=C(C)C1=NC(=CC3=NC(=C(C)C4=NC2(C)C(C)(CC(N)=O)C4CCC(N)=O)C(C)(CC(N)=O)C3CCC(N)=O)C(C)(C)C1CCC(N)=O)OP([O-])(=O)OC1C(CO)OC(C1O)N1C=NC2=CC(C)=C(C)C=C12
Especificaciones
Nombre del producto químico o material Vitamin B12a hydrochloride
Cantidad 1 g
Formula Weight (peso de la fórmula) 1346.35