Learn More
Vimentin Rabbit Recombinant Antibody, Proteintech
Código de producto. 30233894 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
100 μL
20 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
30233894 100 μL 100 microlitros
30236843 20 μL 20 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 30233894 Proveedor Proteintech N.º de proveedor 802321RR100UL
Rabbit Recombinant Antibody
The Vimentin antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Vimentin. This antibody recognizes human, mouse, rat antigen. The Vimentin antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.
Especificaciones
Antígeno Vimentin
Aplicaciones Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA
Clasificación Recombinant Monoclonal
Clon 6K21
Concentración 500 μg/mL
Conjugado Unconjugated
Símbolos de los genes FLJ36605
Especie del huésped Rabbit
Inmunógeno Recombinant Protein
Método de purificación Protein A purification
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 7431
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at -20°C. Stable for one year after shipment.
Peso molecular del anticuerpo 50-55 kDa
Tipo de producto Recombinant Antibody
Formulario Liquid
Isotype IgG