Learn More
VIM Mouse anti-Human, Clone: LN-6, Abnova™
Descripción
This gene encodes a member of the intermediate filament family. Intermediate filamentents, along with microtubules and actin microfilaments, make up the cytoskeleton. The protein encoded by this gene is responsible for maintaining cell shape, integrity of the cytoplasm, and stabilizing cytoskeletal interactions. It is also involved in the immune response, and controls the transport of low-density lipoprotein (LDL)-derived cholesterol from a lysosome to the site of esterification. It functions as an organizer of a number of critical proteins involved in attachment, migration, and cell signaling. Mutations in this gene causes a dominant, pulverulent cataract
Especificaciones
Especificaciones
| Antígeno | VIM |
| Aplicaciones | Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) |
| Clasificación | Monoclonal |
| Clon | LN-6 |
| Conjugado | Unconjugated |
| Descripción | Mouse monoclonal antibody raised against native human VIM. |
| Dilución | Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulación | In 10mM PBS. |
| Alias de gen | FLJ36605 |
| Símbolos de los genes | VIM |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.