Learn More
SMC1 Antibody, CoraFluor™ 1, Novus Biologicals™
Descripción
SMC1 Polyclonal antibody specifically detects SMC1 in Human,Mouse samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Especificaciones
Especificaciones
| Antígeno | SMC1 |
| Aplicaciones | Immunohistochemistry, Immunoprecipitation, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | CoraFluor 1 |
| Formulación | PBS |
| Alias de gen | CDLS2, DXS423ESMC1 structural maintenance of chromosomes 1-like 1 (yeast), KIAA0178DKFZp686L19178, Sb1.8, SB1.8MGC138332, segregation of mitotic chromosomes 1, SMC protein 1A, SMC1 (structural maintenance of chromosomes 1, yeast)-like 1, SMC1 structural maintenance of chromosomes 1-like 1, SMC-1A, SMC-1-alpha, SMC1L1, SMC1SMC1alpha, SMCB, structural maintenance of chromosomes 1A, structural maintenance of chromosomes protein 1A |
| Especie del huésped | Rabbit |
| Inmunógeno | The immunogen this antibody was made to, maps to a region of human SMC1 (structural maintenance of chromosomes 1-like 1 ) surrounding serine 957 using the numbering given in entry NP_006297.2 (Gene ID 8243) when the serine residue is phosphorylated. |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Mostrar más |