Learn More
Human TAP2 Antibody, R&D Systems™
Descripción
TAP2 Monoclonal specifically detects TAP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Especificaciones
Especificaciones
| Antígeno | TAP2 |
| Aplicaciones | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | 998524 |
| Conjugado | Unconjugated |
| Dilución | Western Blot 2 μg/mL, Immunohistochemistry 8-25 μg/mL, Immunocytochemistry 8-25 μg/mL |
| Formulación | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. |
| N.º de referencia del gen | Q03519 |
| Alias de gen | ABC18, ABCB3, ABCB3ABC18, antigen peptide transporter 2, APT2, ATP-binding cassette, sub-family B (MDR/TAP), member 3, D6S217E, MHC 2, Peptide transporter involved in antigen processing 2, Peptide transporter PSF2, Peptide transporter TAP2, PSF2, PSF2PSF-2, Really interesting new gene 11 protein, RING11, RING11ATP-binding cassette sub-family B member 3, transporter 2, ABC (ATP binding cassette), transporter 2, ATP-binding cassette, sub-family B (MDR/TAP), Y1 |
| Especie del huésped | Mouse |
| Mostrar más |