Learn More
Human Cerebellin-2 Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18111631
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18111631 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Código de producto. 18111631 Proveedor R&D Systems N.º de proveedor MAB70441
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Mouse Monoclonal Antibody
Cerebellin-2 Monoclonal specifically detects Cerebellin-2 in Human samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Cerebellin-2
Aplicaciones Immunohistochemistry
Clasificación Monoclonal
Clon 704323
Concentración LYOPH
Conjugado Unconjugated
Dilución Immunohistochemistry 8-25 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen Q8IUK8
Alias de gen CBLN2, cerebellin 2 precursor, cerebellin-2, Cerebellin2
Símbolos de los genes CBLN2
Especie del huésped Mouse
Inmunógeno Chinese hamster ovary cell line CHO-derived recombinant human Cerebellin-2 Arg50-Leu224 Accession # Q8IUK8
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 147381
Especificidad de la prueba Detects human Cerebellin-2 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) Cerebellin-1 and rhCerebellin-4 is observed, and approximately 50% cross-reactivity with rhCerebellin-3 is observed.
Reconstitución Sterile PBS to a final concentration of 0.5 mg/mL.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG1