Learn More
MDK83190, MedChemExpress
Código de producto. 16497548
Encompass_Preferred
Change view
Click to view available options
Cantidad:
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16497548 10 mg 10 miligramos
16427496 100 mg 100 miligramos
16413577 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16497548 Proveedor Medchem Express N.º de proveedor HY18633
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
MedChemExpress MDK83190 is a potent apoptosis activator, induces Apaf-1 oligomerization, increases procaspase-9 processing and subsequent caspase-3 activation in a cyto c-dependent Manner.
MDK83190 is a potent apoptosis activator, induces Apaf-1 oligomerization, increases procaspase-9 processing and subsequent caspase-3 activation in a cyto c-dependent Manner.
Specifications
Nombre del producto químico o material MDK83190
Forma física Solid
CAS 79183-19-0
Color Naranja
Cantidad 10 mg
Notas de grado de pureza Research
Fórmula molecular C15H9Cl2NO2
Para utilizar con (aplicación) Cancer-programmed cell death
Información de solubilidad DMSO : ≥ 46 mg/mL (150.26 mM)
SMILES O=C1N(CC2=CC=C(Cl)C(Cl)=C2)C3=C(C=CC=C3)C1=O
Peso molecular (g/mol) 306.14
Formula Weight (peso de la fórmula) 306.14
Porcentaje de pureza 98.08%
Grado Research
Show More Show Less
Research purposes only
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.