Learn More
CCNC (phospho S275), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Descripción
The protein encoded by this gene is a member of the cyclin family of proteins. The encoded protein interacts with cyclin-dependent kinase 8 and induces the phophorylation of the carboxy-terminal domain of the large subunit of RNA polymerase II. The level of mRNAs for this gene peaks in the G1 phase of the cell cycle. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | CCNC |
| Aplicaciones | Immunohistochemistry (PFA fixed), Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Descripción | Rabbit polyclonal antibody raised against synthetic phosphopeptide of human CCNC. |
| Dilución | Immunohistochemistry (1:50-1:100) Western Blot (1:500-1000) The optimal working dilution should be determined by the end user. |
| Formulación | In PBS, pH 7.4 (without mg2+ and Ca2+), (50% glycerol, 0.02% sodium azide) |
| génica | CCNC |
| N.º de referencia del gen | P24863 |
| Alias de gen | CycC |
| Mostrar más |