Learn More
polymerase (RNA) III (DNA directed) polypeptide C (62kDa), Mouse, Polyclonal Antibody, Abnova™
Descripción
Sequence: MTQAEIKLCSLLLQEHFGEIVEKIGVHLIRTGSQPLRVIAHDTGTSLDQVKKALCVLVQHNLVSYQVHKRGVVEYEAQCSRVLRMLRYPRYIYTTKTLYSDTGELIVEELLLNGKLTMSAVVKKVADRLTETMEDGKTMDYAEVSNTFVRLADTHFVQRCPSVPTTENSDPGPPPPAPTLVINEKDMYLVPKLSLIGKGKRRRSSDEDAAGEPKAKRPKYTTDNKEPIPDDGIYWQANLDRFHQHFRDQAIVSAVANRMDQTSSEIVRTMLRMSEITTSSSAPFTQPLSSNEIFRSLPVGYNISKQVLDQYLTLLADDPLEFVGKSGDSGGGMYVINLHKALASLATATLESVVQERFGSRCARIFRLVLQKKHIEQKQVEDFAMIPAKEAKDMLYKMLSENFMSLQEIPKTPDHAPSRTFYLYTVNILSAARMLLHRCYKSIANLIERRQFETKENKRLLEKSQRVEAIIASMQATGAEEAQLQEIEEMITAPERQQLETLKRNVNKLDASEIQVDETIFLLESYIECTMKRQ
Especificaciones
Especificaciones
| Antígeno | polymerase (RNA) III (DNA directed) polypeptide C (62kD) |
| Aplicaciones | Immunofluorescence, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Descripción | Mouse polyclonal antibody raised against a full-length human POLR3C protein. |
| Formulación | No additive |
| génica | POLR3C |
| N.º de referencia del gen | NM_006468.5 |
| Alias de gen | RPC3/RPC62 |
| Símbolos de los genes | POLR3C |
| Mostrar más |