Learn More
APOL4, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Descripción
The protein encoded by this gene is a member of the apolipoprotein L family and may play a role in lipid exchange and transport throughout the body, as well as in reverse cholesterol transport from peripheral cells to the liver. Two transcript variants encoding two different isoforms have been found for this gene. Only one of the isoforms appears to be a secreted protein. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | APOL4 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Descripción | Mouse polyclonal antibody raised against a full-length human APOL4 protein. |
| Formulación | PBS with no preservative; pH 7.4 |
| génica | APOL4 |
| N.º de referencia del gen | ENST00000328429 |
| Alias de gen | APOL-IV/APOLIV |
| Símbolos de los genes | APOL4 |
| Mostrar más |