Learn More
Rho guanine nucleotide exchange factor (GEF) 5, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Código de producto. 16168105
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16168105 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Código de producto. 16168105 Proveedor Abnova N.º de proveedor H00007984D01P.100ug
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Rabbit polyclonal antibody raised against a full-length human ARHGEF5 protein.
Rho GTPases play a fundamental role in numerous cellular processes initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form a complex with G proteins and stimulate Rho-dependent signals. This protein may be involved in the control of cytoskeletal organization. [provided by RefSeq
Sequence: MGGFSRRCSKLINSSQLLYQEYSDVVLNKEIQSQQRLESLSETPGPSSPRQPRKALVSSESYLQRLSMASSGSLWQEIPVVRNSTVLLSMTHEDQKLQEVKFELIVSEASYLRSLNIAVDHFQLSTSLRATLSNQEHQWLFSRLQDVRDVSATFLSDLEENFENNIFSFQVCDVVLNHAPDFRRVYLPYVTNQTYQERTFQSLMNSNSNFREVLEKLESDPVCQRLSLKSFLILPFQRITRLKLLLQNILKRTQPGSSEEAEATKAHHALEQLIRDCNNNVQSMRRTEELIYLSQKIEFECKIFPLISQSRWLVKSGELTALEFSASPGLRRKLNTRPVHLHLFNDCLLLSRPREGSRFLVFDHAPFSSIRGEKCEMKLHGPHKNLFRLFLRQNTQGAQAEFLFRTETQSEKLRWISALAMPREELDLLECYNSPQVQCLRAYKPRENDELALEKADVVMVTQQSSDGWLEGVRLSDGERGWFPVQQVEFISNPEVRAQNLKEAHRVKTAKLQLVEQQA
Especificaciones
Antígeno Rho guanine nucleotide exchange factor (GEF) 5
Aplicaciones Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Descripción Rabbit polyclonal antibody raised against a full-length human ARHGEF5 protein.
Formulación PBS with no preservative; pH 7.4
génica ARHGEF5
N.º de referencia del gen Q12774
Alias de gen DKFZp686N1969/GEF5/P60/TIM/TIM1
Símbolos de los genes ARHGEF5
Especie del huésped Rabbit
Inmunógeno ARHGEF5 (Q12774, 1 a.a. ∼ 519 a.a) full-length human protein.
Método de purificación Affinity Purified
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 7984
Especies diana Human
Contenido y almacenamiento Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Tipo de producto Antibody
Isotype IgG