Learn More
VH 032 - linker 3, Tocris Biosciences™
PROTACs (Proteolysis Targeting Chimeras)
Marca: Tocris Bioscience 6465/25
Descripción
VH 032 amide-alkylC4-amine is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC® research and development; incorporates an E3 ligase ligand plus a short alkyl linker ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.This product has been recently renamed. The previous name for this product was VH 032 - linker 3
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Especificaciones
| PROTAC Toolbox | |
| 2341796-78-7 | |
| VH 032 - linker 3 | |
| 25 mg |