Learn More
Tocris Bioscience™ VH 032 amide-alkylC8-acid
Descripción
VH 032 amide-alkylC8-acid is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC® research and development; incorporates an E3 ligase ligand plus alkylC8 linker with terminal carboxylic acid ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D. PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 10-((-1-((2 S,4 R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-10-oxodecanoic acid |
| CAS | 2172819-77-9 |
| Cantidad | 25 mg |
| Objetivo | Degrader Building Blocks |
| Fórmula molecular | C32H46N4O6S |
| Pureza | 0.95 |