Learn More
Valnemulin hydrochloride, MedChemExpress
Código de producto. 16468066
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16468066 100 mg 100 miligramos
16448328 10 mg 10 miligramos
16404217 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16468066 Proveedor Medchem Express N.º de proveedor HYB0027
MedChemExpress Valnemulin hydrochloride is a pleuromutilin antibiotic which inhibits protein synthesis in bacteria by binding the peptidyl transferase enzyme in the 50s ribosomal subunit.
Valnemulin hydrochloride is a pleuromutilin antibiotic which inhibits protein synthesis in bacteria by binding the peptidyl transferase enzyme in the 50s ribosomal subunit.
Especificaciones
Nombre del producto químico o material Valnemulin hydrochloride
Forma física Solid
CAS 133868-46-9
Color Blanco
Cantidad 100 mg
Notas de grado de pureza Research
Fórmula molecular C31H53ClN2O5S
Información de solubilidad DMSO : ≥ 100 mg/mL (166.31 mM) ∣H2O : 100 mg/mL (166.31 mM; Need ultrasonic)
SMILES C[C@@H]([C@@H]([C@@](C)(C[C@H]1OC(CSC(C)(C)CNC([C@H](N)C(C)C)=O)=O)C=C)O)C(CCC2=O)(CC[C@H]3C)[C@]2([H])C31C.Cl
Peso molecular (g/mol) 601.28
Formula Weight (peso de la fórmula) 601.28
Porcentaje de pureza 98.3%