Descripción
V-801 EasyVac
EasyVac system is a dedicated module designed to work specifically with the V-700 or V-710 pumps to allow automatic distillations with any rotary evaporator. The module gradually lowers the vacuum until solvent vapour is detected. Once all vapour has been evaporated, the controller then looks for other solvents. The process is repeated until all solvents have been distilled off.
V-850 controller
Semi-automatic controller. With solvent library of 43 common solvents and timer function. Full manual control also possible, including vacuum release.
V-855 controller
Automatic controller. As V-850 plus automatic distillation functions based on either vapour pressure or vapour temperature (latter requires optional autodistillation probe).
EasyVac system is a dedicated module designed to work specifically with the V-700 or V-710 pumps to allow automatic distillations with any rotary evaporator. The module gradually lowers the vacuum until solvent vapour is detected. Once all vapour has been evaporated, the controller then looks for other solvents. The process is repeated until all solvents have been distilled off.
V-850 controller
Semi-automatic controller. With solvent library of 43 common solvents and timer function. Full manual control also possible, including vacuum release.
V-855 controller
Automatic controller. As V-850 plus automatic distillation functions based on either vapour pressure or vapour temperature (latter requires optional autodistillation probe).
- Vacuum controllers are multi-function units with a bright LCD digital and analogue graphics display
- Range of pre-loaded applications
- All units fully compatible with Büchi and other brands of vacuum pumps
- Retort stand mounting
Especificaciones
Especificaciones
| Tipo | Vacuum Controller V-855 |
| Cantidad | 1 unidad |
| Requisitos eléctricos | 85/264 V 50/60 Hz |
| Tensión | 85/264 V |
| Frecuencia | 50/60 Hz |