Learn More
MedChemExpress Ustekinumab
Specifications
Specifications
| Antígeno | Interleukin Related |
| Cantidad | 5 mg |
| Especies diana | Human |
| Contenido y almacenamiento | Please store the product under the recommended conditions in the Certificate of Analysis. |
| Tipo de producto | Inhibitory Antibodies |
| Condiciones de envío | Shipping with dry ice |
| CAS | 815610-63-0 |
| Grado de pureza o calidad | 98.42% |