missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UNC5CL Polyclonal Antibody

Descripción
This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
UNC5CL is a membrane protein belonging to theUNC-5 family with a death domain and a ZU5 domain and is known to interact with p65/RELA and NFKB1. UNC5CL is known to interact with subunit p105 of NF-kappaB and transactivator p65 and is thus involved in inhibition of NF-kappaB activation and NF-kappaB-dependent transcription by inhibiting the binding of NF-kappaB to its target sequence (providing an alternative regulatory mechanism for NF-kappaB-mediated transcription). This regulatory protein sensitizes cells to apoptosis induced by TNF and TRAIL.
Especificaciones
Especificaciones
| Antígeno | UNC5CL |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| génica | UNC5CL |
| N.º de referencia del gen | Q8IV45 |
| Alias de gen | 2510009H09Rik; BB155270; MGC34763; MUXA; OTTHUMP00000016362; protein unc-5 homolog C-like; unc-5 family C-terminal like; unc-5 homolog C (C. elegans)-like; UNC5CL; UNC5C-like protein; zu-5; ZU5 and death domain containing; ZU5 and death domain-containing inhibitor of NF-kB; ZU5 and death domain-containing protein; ZUD |
| Símbolos de los genes | UNC5CL |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?