Learn More
UNC0379, MedChemExpress
Código de producto. 16485098
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16485098 10 mg 10 miligramos
16455826 100 mg 100 miligramos
16451337 50 mg 50 miligramos
16417827 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16485098 Proveedor Medchem Express N.º de proveedor HY12335
MedChemExpress UNC0379 is a selective, substrate-competitive inhibitor of lysine methyltransferase SETD8 (KMT5A) with an IC50 of 7.3 μM, KD value of 18.3 μM. UNC0379 can be used in the research of inflammation and cancers, such as pulmonary fibrosis, ovarian cancer, neuroblastoma.
UNC0379 is a selective, substrate-competitive inhibitor of lysine methyltransferase SETD8 (KMT5A) with an IC50 of 7.3 μM, KD value of 18.3 μM. UNC0379 can be used in the research of inflammation and cancers, such as pulmonary fibrosis, ovarian cancer, neuroblastoma.
Especificaciones
Nombre del producto químico o material UNC0379
CAS 1620401-82-2
Color Earth Yellow
Notas de grado de pureza Research
Fórmula molecular C23H35N5O2
Cantidad 10 mg
Información de solubilidad DMSO : 50 mg/mL (120.90 mM; Need ultrasonic)
SMILES COC1=C(OC)C=C2C(N=C(N3CCCC3)N=C2NCCCCCN4CCCC4)=C1
Peso molecular (g/mol) 413.56
Formula Weight (peso de la fórmula) 413.56
Porcentaje de pureza 99.53%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death