Learn More
STEMCELL Technologies UM729
Código de producto. 17109251
Encompass_Preferred
Change view
Click to view available options
Cantidad:
250 μg de componente activo
Tamaño de la unidad:
250 microgramos
17109251 250 μg de componente activo 250 microgramos
1 options
Código de producto. 17109251
Marca: STEMCELL Technologies 72332
Pyrimido-indole derivative that enhances HSC self-renewal in vitro
  • UM729 is a pyrimido-[4,5-b]-indole derivative which enhances the self-renewal of human hematopoietic stem cells in vitro
  • UM729 does not inhibit the aryl hydrocarbon receptor (AHR) pathway, but has been shown to collaborate with AHR antagonists in preventing differentiation of acute myeloid leukemia (AML) cells in culture
  • Enhances human hematopoietic stem cell self-renewal in vitro (Fares et al.)
  • Collaborates with StemRegenin 1 (SR1) in preventing differentiation of AML cells in culture (Pabst et al.)
Especificaciones
Sinónimo UM-729
Cantidad 250 μg de componente activo
Fórmula molecular C20H25N5O2 · X HCl [X H2O]
Pureza ≥95%