Learn More
Dr. Ehrenstorfer Inspección de Agua Potable del Reino Unido (DWI) Kit etiquetado PFAS-35 2 μg/mL en Metanol:Agua
Descripción
- Materiales de referencia certificados
Especificaciones
Especificaciones
| Nombre del producto químico o material | N-Ethyl-N-(2-hydroxyethyl)perfluorooctylsulphonamide-d9, Perfluoro-2-propoxypropanoic Acid 13C3 (Propionic Acid 13C3), Perfluorooctanesulfonic acid 13C8, Perfluoroundecanoic Acid 13C7 (1,2,3,4,5,6,7-13C7), 2-(N-Methylperfluorooctanesulfonamido)acetic acid D3 (N-methyl D3), N-Methylperfluorooctanesulfonamidoethanol-d7, 1H,1H,2H,2H-Perfluorooctanesulfonic Acid 13C2 (1,2-13C2), 2-(N-Ethylperfluorooctanesulfonamido)acetic Acid D5 (N-Ethyl D5), Perfluorohexanoic Acid 13C5 (1,2,3,4,6-13C5), N-Methylperfluorooctanesulfonamide-d3, Perfluorooctanoic acid 13C8, Capstone Product B 13C3 (Dimethyl 13C2, Glycine-beta-13C1), Perfluorobutanoic acid 13C4, Perfluoroheptanoic Acid 13C4 (1,2,3,4-13C4), Perfluorononanoic acid 13C9, Perfluorotetradecanoic Acid 13C2 (1,2-13C2), 1H,1H,2H,2H-Perfluorodecanesulfonic Acid 13C2 (1,2-13C2), Perfluorohexadecanoic acid 13C2 (1,2-13C2), Perfluorotridecanoic acid 13C2 (1,2-13C2), Perfluorooctane Sulfonamide 13C8, 1H,1H,2H,2H-Perfluorohexanesulfonic Acid 13C2 (1,2-13C2), Perfluoro-4-methoxybutanoic Acid 13C2 (1,2-13C2), Perfluoropentanoic acid 13C5, Perfluorodecanoic acid 13C6 (1,2,3,4,5,6-13C6), Perfluorobutanesulfonic Acid 13C3 (2,3,4-13C3), Perfluorohexanesulfonic Acid 13C3 (1,2,3-13C3), Sulfluramid ethyl-d5, Perfluorododecanoic Acid (1,2-13C2), Perfluorodecanesulfonic Acid 13C3 (1,2,3-13C3), Perfluorododecanesulfonic Acid 13C3 (1,2,3-13C3), Perfluoroheptanesulfonic Acid 13C3 (1,2,3-13C3), Perfluorononanesulfonic Acid 13C3 (1,2,3-13C3), Perfluoropentanesulfonic Acid 13C3 (1,2,3-13C3), Perfluorotridecanesulfonic Acid 13C3 (1,2,3-13C3), Perfluoroundecanesulfonic Acid 13C3 (1,2,3-13C3) |
| CAS | 1265205-96-6, 3030247-97-0, 2265893-05-6, 1400690-70-1, 1265205-95-5, 2088416-87-7 |
| Disolvente o matriz | Methanol/Water |
| Cantidad | 2 x 1 mL |