Learn More
Tyk2-IN-5, MedChemExpress
Código de producto. 16485376
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
25 miligramos
5 miligramos
50 miligramos
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
16485376 100 mg 100 miligramos
16484518 10 mg 10 miligramos
16453116 25 mg 25 miligramos
16450827 50 mg 50 miligramos
16417237 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Código de producto. 16485376 Proveedor Medchem Express N.º de proveedor HY111745
MedChemExpress Tyk2-IN-5 (compound 6) is a highly potent, selective and orally active Tyk2 inhibitor and targets the JH2 domain, with a Ki of 0.086 nM for Tyk2 JH2 and an IC50 of 25 nM for IFNα. Highly effective in inhibiting IFNγ production in a rat pharmacodynamics model and fully efficacious in a rat adjuvant arthritis model.
Tyk2-IN-5 (compound 6) is a highly potent, selective and orally active Tyk2 inhibitor and targets the JH2 domain, with a Ki of 0.086 nM for Tyk2 JH2 and an IC50 of 25 nM for IFNα. Highly effective in inhibiting IFNγ production in a rat pharmacodynamics model and fully efficacious in a rat adjuvant arthritis model.
Especificaciones
Nombre del producto químico o material Tyk2-IN-5
CAS 1797432-62-2
Color Light Green
Notas de grado de pureza Research
Fórmula molecular C21H19FN8O2
Cantidad 100 mg
Información de solubilidad DMSO : 43.33 mg/mL (99.74 mM; Need ultrasonic)
SMILES O=C1C(NC2=NN3C(C(NC)=C2)=NC=C3C(N[C@H]4[C@@H](F)C4)=O)=CC=CN1C5=CC=CC=N5
Peso molecular (g/mol) 434.43
Formula Weight (peso de la fórmula) 434.43
Porcentaje de pureza 98.87%
Grado Research
Forma física Solid
Para utilizar con (aplicación) COVID-19-immunoregulation