Learn More
Tocris Bioscience™ Tucatinib
Descripción
Tucatinib is a potent and selective ErbB-2 (also called HER2) kinase inhibitor (IC50 = 7 nM). It exhibits >50-fold selectivity for ErbB-2 over EGFR. In vitro, Tucatinib inhibits ErbB-2 signaling activity and cell proliferation in ErbB-2-amplified breast cancer cell lines (BT-474). In vivo, Tucatinib (25-100 mg/kg) reduces tumor volume in breast and gastric xenograft mouse models.This compound is orally bioavailable.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N6-(4,5-Dihydro-4,4-dimethyl-2-oxazolyl)-N4-[3-methyl-4-([1,2,4 ]triazolo[1,5-a]pyridin-7-yloxy)phenyl]-4,6-quinazolinediamine |
| CAS | 937263-43-9 |
| Cantidad | 10 mg |
| Objetivo | EGF Receptor Inhibitors |
| Fórmula molecular | C26H24N8O2 |
| Pureza | 0.98 |