Learn More
Tubulin inhibitor 1, MedChemExpress
Código de producto. 16460947
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16460947 50 mg 50 miligramos
16454658 10 mg 10 miligramos
16427377 5 mg 5 miligramos
16425496 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16460947 Proveedor Medchem Express N.º de proveedor HY112607
This item is not returnable. View return policy
MedChemExpress Tubulin inhibitor 1 is a tubulin inhibitor, inhibits tubulin polymerization. Tubulin inhibitor 1 shows potent anti-tumor activity, casues cellular mitotic arrest in the G2/M phase, and induces cellular apoptosis.
Tubulin inhibitor 1 is a tubulin inhibitor, inhibits tubulin polymerization. Tubulin inhibitor 1 shows potent anti-tumor activity, casues cellular mitotic arrest in the G2/M phase, and induces cellular apoptosis.
Specifications
Nombre del producto químico o material Tubulin inhibitor 1
CAS 2237054-53-2
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C21H24N2O4
Cantidad 50 mg
Información de solubilidad DMSO : ≥ 125 mg/mL (339.28 mM)
SMILES CCOC1=CC=C(C2=CN(C)N=C2C3=CC(OC)=C(OC)C(OC)=C3)C=C1
Peso molecular (g/mol) 368.43
Formula Weight (peso de la fórmula) 368.43
Porcentaje de pureza 99.67%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Neuroscience-Neurodegeneration