Learn More
Tocris Bioscience™ Tubastatin A hydrochloride
Descripción
Tubastatin A hydrochloride is a potent HDAC6 inhibitor (IC50 = 0.015 μM). Exhibits some selectively for HDAC6 over HDAC8 and 1 (IC50 values are 0.854 and 16.4 μM, respectively). Induces elevated levels of α-tubulin and protects against glutathione-induced oxidative stress in primary neuronal cell culture. Reverses the axonal loss in peripheral neurons in a mouse model of Charcot-Marie-Tooth disease.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N-Hydroxy-4-[(1,2,3,4-tetrahydro-2-methyl-5H-pyrido[4,3-b]indol-5-yl)methyl]benzamide hydrochloride |
| CAS | 1310693-92-5 |
| Intervalo de porcentaje de ensayo | >98% |
| Cantidad | 10 mg |
| Fórmula molecular | C20H21N3O2.HCl |
| Formula Weight (peso de la fórmula) | Observed MW: 371.86 |