missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSSC1 Polyclonal Antibody

Descripción
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (90%), Rat (90%), Zebrafish (83%), Xenopus laevis (82%), Pig (83%), Chicken (92%), Bovine (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TSSC1 has been reported in PMID 9403053 as one of several tumor-suppressing sub transferable fragments located in the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. Alignment of this gene to genomic sequence data suggests that this gene resides on chromosome 2 rather than chromosome 11.
Especificaciones
Especificaciones
| Antígeno | TSSC1 |
| Aplicaciones | Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.75 mg/mL |
| Conjugado | Unconjugated |
| Formulación | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| génica | EIPR1 |
| N.º de referencia del gen | Q53HC9 |
| Alias de gen | EARP and GARP complex-interacting protein 1; EIPR1; Endosome-associated recycling protein-interacting protein; Golgi-associated retrograde protein-interacting protein; protein TSSC1; Tssc1; tumor suppressing subtransferable candidate 1; tumor-suppressing STF cDNA 1 protein; Tumor-suppressing subchromosomal transferable fragment candidate gene 1 protein |
| Símbolos de los genes | EIPR1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?