missing translation for 'onlineSavingsMsg'
Learn More

TRPV1 Polyclonal Antibody, Invitrogen™

Código de producto. 11552332
Cambiar vista
Click to view available options
Cantidad:
50 μL
Tamaño de la unidad:
50 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11552332 50 μL 50 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11552332 Proveedor Invitrogen N.º de proveedor PA129421

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

PA1-29421 detects Vanilloid receptor 1 from rat, human, mouse, and non-human primate samples. PA1-29421 has been successfully used in immunocytochemistry, immunohistochemistry (frozen tissue), and Western blot applications. A suggested positive control for this product is VR1 transfected cells. Predicted molecular weight of approximately 100.7 kDa. The PA1-29421 immunogen is synthetic peptide, RASLDSEESESPPQENSC, corresponding to amino-terminus of Rat VR1, amino acids 4-21. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

TRPV1 (Transient Receptor Potential Cation Channel Subfamily V Member 1) is a receptor for capsaicin, the main pungent ingredient in hot chili peppers, and elicits a sensation of burning pain by selectively activating sensory neurons. TRPV1 is a non-selective cation channel that is structurally related to members of the TRP family of ion channels such as TRPC3 and TRPC6. TRPV1 is permeable to calcium, magnesium, and monovalent cations of sodium and potassium. Structurally, TRPV1 contains 6 trans-membrane domains, a P-loop membrane domain and 3 ankyrin repeat domains. TRPV1 is also activated by increases in temperature in the noxious range, suggesting that it functions as a transducer of painful thermal stimuli in vivo. TRPV1 seems to mediate proton influx and may be involved in intracellular acidosis in nociceptive neurons. The expression of TRPV1 appears to be limited to neurons within the sensory ganglia. Four transcript variants encoding TRPV1 with variations in th 5′ UTR sequence have been described. Diseases associated with TRPV1 include Irritable Bowel Syndrome (IBS) and Bladder Disease.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno TRPV1
Aplicaciones Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
génica TRPV1
N.º de referencia del gen Q8NER1, Q704Y3, O35433
Alias de gen VR1, Transient receptor potential cation channel subfamily V member 1, TrpV1, osm-9-like TRP channel 1, OTRPC1, Capsaicin Receptor, Vanilloid receptor
Símbolos de los genes TRPV1
Especie del huésped Rabbit
Inmunógeno synthetic peptide, RASLDSEESESPPQENSC, corresponding to amino-terminus of Rat VR1, amino acids 4-21.
Cantidad 50 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 193034, 7442, 83810
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.