Learn More
Tocris Bioscience™ TRPC6-PAM-C20
Descripción
TRPC6-PAM-C20 is a selective TRPC6 positive allosteric modulator. Selectively activates TRPC6 over other TRP channels. Also activates TRPA1. Induces transient increase in intracellular Ca2+ in HEK cells expressing TRPC6 (EC50 = 2.37 μM). Potentiates OAG (1-oleoyl-1-acetyl-sn-glycerol)-induced Ca2+ influx in human platelets, and enhances OAG-induced platelet aggregation.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 3-(3-,4-Dihydro-6,7-dimethoxy-3,3-dimethyl-1-isoquinolinyl)-2H-1-benzopyran-2-one |
| CAS | 667427-75-0 |
| Cantidad | 10 mg |
| Fórmula molecular | C22H21NO4 |
| Pureza | 98% |