Learn More
Fosfato de tris(1,3-dicloro-2-propilo)-d15, TRC
Product Code. 30454864
Change view
Click to view available options
Cantidad:
2.5 mg
Unit Size:
2.50 miligramo
Product Code. Cantidad unitSize
30454864 2.5 mg 2.50 miligramo
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30454864 Supplier Toronto Research Chemicals Supplier No. TRCT8763072.5MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Moléculas orgánicas de alta pureza y estándares analíticos, entregados estratégicamente en todo el mundo para potenciar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material Tris(1,3-dichloro-2-propyl) phosphate-d15
CAS 1447569-77-8
Notas de grado de pureza HPLC
Fórmula molecular C9 D15 Cl6 O4 P
Cantidad 2.5 mg
Sinónimo Tris(2-chloro-1-(chloromethyl)ethyl) phosphate D15, Tris(1,3-dichloro-2-propyl) phosphate D15, Tris(1,3-dichloroisopropyl) phosphate D15
Fórmula InChI InChI=1S/C9H15Cl6O4P/c10-1-7(2-11)17-20(16,18-8(3-12)4-13)19-9(5-14)6-15/h7-9H,1-6H2/i1D2,2D2,3D2,4D2,5D2,6D2,7D,8D,9D
SMILES [2H]C([2H])(Cl)C([2H])(OP(=O)(OC([2H])(C([2H])([2H])Cl)C([2H])([2H])Cl)OC([2H])(C([2H])([2H])Cl)C([2H])([2H])Cl)C([2H])([2H])Cl
Nombre IUPAC fosfato de tris[2-cloro-1-[cloro(dideuterio)metil]-1,2,2-trideuterio-etilo]
Peso molecular (g/mol) 446
Formula Weight (peso de la fórmula) 442.978
Porcentaje de pureza >95