missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIP12 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15765731 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15765731 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Artikelnummer. 15765731 Leverandør Invitrogen™ Leverandørnr. PA557840

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

Immunogen sequence: AMCKEIIPTS EFINSKLTAK ANRQLQDPLV IMTGNIPTWL TELGKTCPFF FPFDTRQMLF YVTAFDRDRA MQRLLDTNPE INQSDSQD Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

E3 ubiquitin-protein ligase involved in ubiquitin fusion degradation (UFD) pathway and regulation of DNA repair. Part of the ubiquitin fusion degradation (UFD) pathway, a process that mediates ubiquitination of protein at their N-terminus, regardeless of the presence of lysine residues in target proteins. In normal cells, mediates ubiquitination and degradation of isoform p19ARF/ARF of CDKN2A, a lysine-less tumor suppressor required for p53/TP53 activation under oncogenic stress. In cancer cells, however, isoform p19ARF/ARF and TRIP12 are located in different cell compartments, preventing isoform p19ARF/ARF ubiquitination and degradation. Does not mediate ubiquitination of isoform p16-INK4a of CDKN2A. Also catalyzes ubiquitination of NAE1 and SMARCE1, leading to their degradation. Ubiquitination and degradation of target proteins is regulated by interaction with proteins such as MYC, TRADD or SMARCC1, which disrupt the interaction between TRIP12 and target proteins. Acts as a key regulator of DNA damage response by acting as a suppressor of RNF168, an E3 ubiquitin-protein ligase that promotes accumulation of 'Lys-63'-linked histone H2A and H2AX at DNA damage sites, thereby acting as a guard against excessive spreading of ubiquitinated chromatin at damaged chromosomes.
TRUSTED_SUSTAINABILITY

Tekniske data

Antígeno TRIP12
Aplicaciones Immunohistochemistry (Paraffin), Immunocytochemistry
Clasificación Polyclonal
Concentración 0.2 mg/mL
Conjugado Unconjugated
Formulación PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
génica TRIP12
N.º de referencia del gen Q14669
Alias de gen 1110036I07Rik; 6720416K24Rik; AA410158; E3 ubiquitin-protein ligase for Arf; E3 ubiquitin-protein ligase TRIP12; gene trap locus 6; Gtl6; HECT-type E3 ubiquitin transferase TRIP12; KIAA0045; probable E3 ubiquitin-protein ligase TRIP12; thyroid hormone receptor interactor 12; thyroid receptor interacting protein 12; thyroid receptor interactor TRIP12; thyroid receptor-interacting protein 12; TR-interacting protein 12; TRIP12; TRIP-12; ULF
Símbolos de los genes TRIP12
Especie del huésped Rabbit
Inmunógeno Recombinant protein corresponding to Human TRIP12. Recombinant protein control fragment (Product #RP-96151).
Método de purificación Antigen Affinity Chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 9320
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.