missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIM34 Monoclonal Antibody (K24023_20A3), Invitrogen™

Código de producto. 30195575
Change view
Click to view available options
Cantidad:
50 μL
200 μL
Tamaño de la unidad:
200 microgramos
50 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30195575 50 μL 50 microgramos
30211672 200 μL 200 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30195575 Proveedor Invitrogen™ N.º de proveedor MA555810

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Sequence of this protein is as follows: GEKLLLFCKE DRKVICWLCE RSQEHRGHHT VLTEEVFKEC QEKLQAVLKR LKKEEEEAEK LEADIREEKT SWKYQVQTER QRIQTEFDQL RSILNNEEQR ELQRLEEEEK KTLDKFAEAE

TRIM34 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. Expression of this gene is up-regulated by interferon. This gene is mappped to chromosome 11p15, where it resides within a TRIM gene cluster. Alternate splicing of this gene generates four transcript variants. Additionally, a read-through transcript transcribed from this gene and TRIM6 has been observed.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno TRIM34
Aplicaciones Immunoprecipitation, Western Blot
Clasificación Monoclonal
Clon K24023_20A3
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
génica TRIM34
N.º de referencia del gen Q9BYJ4
Alias de gen IFP1; Interferon-responsive finger protein 1; RING finger protein 21; ring finger protein 21, interferon-responsive; RNF21; TRIM34; tripartite motif containing 34; tripartite motif-containing 34; tripartite motif-containing protein 34
Símbolos de los genes TRIM34
Especie del huésped Mouse
Inmunógeno Recombinant human TRIM34 (immunogen range is 101-220).
Método de purificación Protein A
Cantidad 50 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 53840
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Mostrar más Mostrar menos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.