missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRF1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 11315983
Change view
Click to view available options
Cantidad:
100 μg
Unit Size:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
11315983 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11315983 Supplier Invitrogen™ Supplier No. PA518010

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is predicted to react with bovine and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.

This gene encodes a telomere specific protein which is a component of the telomere nucleoprotein complex. This protein is present at telomeres throughout the cell cycle and functions as an inhibitor of telomerase, acting in cis to limit the elongation of individual chromosome ends. The protein structure contains a C-terminal Myb motif, a dimerization domain near its N-terminus and an acidic N-terminus. Two transcripts of this gene are alternatively spliced products.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno TRF1
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
génica TERF1
N.º de referencia del gen P54274
Alias de gen FLJ41416; hTRF1-AS; NIMA-interacting protein 2; PIN2; Telomeric protein Pin2/TRF1; telomeric repeat binding factor (NIMA-interacting) 1; telomeric repeat binding factor 1; telomeric repeat-binding factor 1; TERF1; TRBF1; TRF; TRF1; TTAGGG repeat-binding factor 1; t-TRF1
Símbolos de los genes TERF1
Especie del huésped Goat
Inmunógeno Synthetic peptide sequence (TMKKLKLISSDSED) corresponding to the C-terminus amino acids of TERF1 (aa 426-439).
Método de purificación Ammonium sulfate precipitation
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 7013
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.