Learn More
Trazodone hydrochloride, Tocris Bioscience™
5-HT2A and α1 adrenoceptor antagonist; also enhances neural differentiation; antidepressant and neuroprotectant
Marca: Tocris Bioscience 6336/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Trazodone hydrochloride is a 5-HT2A and α1 adrenoceptor antagonist. Enhances neuronal differentiation of mouse and human neural progenitor cells. Also inhibits PERK/eIF2α-P-mediated reduction in protein synthesis and restores memory deficits in a mouse dementia model. Antidepressant and neuroprotectant.Especificaciones
| 2-[3-[4-(3-Chlorophenyl)-1-piperazinyl]propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one hydrochloride | |
| 25332-39-2 | |
| >99% | |
| 50 mg |