Learn More
Trazodone hydrochloride, 98+%
Código de producto. p-7042592
Cambiar vista
Click to view available options
Cantidad:
1 g
5 g
Tamaño de la unidad:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad Tamaño de la unidad
15445339 1 g 1 g
15455339 5 g 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 15445339 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor J63070.03
Añadir a la cesta
A serotonin uptake inhibitor | CAS: 25332-39-2 | C19H23Cl2N5O | 408.327 g/mol
A serotonin uptake inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS 25332-39-2
Fórmula molecular C19H23Cl2N5O
Peso molecular (g/mol) 408.327
Número MDL MFCD00079603
Clave InChI OHHDIOKRWWOXMT-UHFFFAOYSA-N
Sinónimo trazodone hydrochloride, trazodone hcl, desyrel, bimaran, molipaxin, devidon, pragmazone, thombran, tombran, tritico
PubChem CID 62935
ChEBI CHEBI:9655
Nombre IUPAC 2-[3-[4-(3-chlorophenyl)piperazin-1-yl]propyl]-[1,2,4]triazolo[4,3-a]pyridin-3-one;hydrochloride
SMILES C1CN(CCN1CCCN2C(=O)N3C=CC=CC3=N2)C4=CC(=CC=C4)Cl.Cl
Especificaciones
Nombre del producto químico o material Trazodone hydrochloride
Punto de fusión 222°C to 227°C
Color Blanco
Cantidad 1 g
Índice Merck 14,9579
Información de solubilidad Soluble at 25mg/ml in methanol; Also soluble in 0;1M HCl or DMSO; Insoluble in water /n
Formula Weight (peso de la fórmula) 408.33
Forma física Powder