Learn More
TRA-1-81 Mouse, anti-Human, APC, Clone: TRA-1-81, STEMCELL Technologies
Código de producto. 17108811
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
100 Tests
25 Tests
Tamaño de la unidad:
100 tests
25 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad Tamaño de la unidad
17108811 100 Tests 100 tests
17118811 25 Tests 25 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 17108811 Proveedor Stemcell Technologies N.º de proveedor 60065AZ
 Artículo restringido
Fecha estimada de envoi: 04-12-2026
Mouse Monoclonal Antibody
  • The TRA-1-81 antibody reacts with a carbohydrate epitope on TRA-1-81, a high-molecular-mass (> 200 kDa) glycoprotein expressed on the surface of human embryonal carcinoma (EC), embryonic germ (EG), embryonic stem (ES), and induced pluripotent stem (iPS) cells, as well as rhesus monkey and rat ES cell lines
  • No immunoreactivity is observed with mouse EC, EG, ES, or iPS cells
  • The epitope of the TRA-1-81 antibody is associated with a keratan-sulfated transmembrane protein identified as podocalyxin and is resistant to treatment with neuraminidase
Especificaciones
Antígeno TRA-1-81
Aplicaciones Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry
Clasificación Monoclonal
Clon TRA-1-81
Conjugado APC
Especie del huésped Mouse
Cantidad 100 Tests
Primario o secundario Primary
Especies diana Rhesus Monkey, Human, Rat
Isotype IgM κ
Tipo de célula Pluripotent Stem Cells